Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176854.2 | NP_789824.1 | 2 - 3 | 376 | 148 | |
GenBank mRNA | BC102897.1 | - | 2 - 3 | 374 | 148 | |
DV837866.1 | - | 2 - 3 | 376 | 148 | ||
M25645.1 | - | 2 - 3 | 373 | 148 | ||
V00113.1 | - | 373 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |