Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177508.1 | NP_803474.1 | 2 - 3 | 343 | 86 | |
GenBank mRNA | BC112872.1 | - | 2 - 3 | 341 | 86 | |
X65125.1 | - | 343 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |