Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164321.1 | NP_001157793.1 | 1 - 2 | 210 | 82 | |
NM_181007.2 | NP_851350.2 | 1 - 2 | 210 | 82 | ||
GenBank mRNA | AB099063.1 | - | 1 - 2 | 146 | 82 | |
BC149388.1 | - | 1 - 2 | 136 | 82 | ||
EE252986.1 | - | 1 - 2 | 231 | 82 | ||
EV793172.1 | - | 1 - 2 | 150 | 82 | ||
J02840.1 | - | 1 - 2 | 133 | 82 | ||
M33268.1 | - | 1 - 2 | 152 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |