Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181010.2 | NP_851353.1 | 1 - 2 | 356 | 97 | |
GenBank mRNA | BC102459.1 | - | 1 - 2 | 376 | 97 | |
S37093.1 | - | 1 - 2 | 356 | 97 | ||
X52740.1 | - | 106 | 97 | |||
X52942.1 | - | 218 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |