Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181031.2 | NP_851374.1 | 2 - 3 | 440 | 81 | |
GenBank mRNA | BC147979.1 | - | 2 - 3 | 529 | 81 | |
BT021727.1 | - | 2 - 3 | 319 | 81 | ||
M16968.1 | - | 2 - 3 | 418 | 81 | ||
X52554.1 | - | 440 | 81 | |||
X72858.1 | - | 478 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |