Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203361.1 | NP_976237.1 | 3 - 4 | 342 | 79 | |
GenBank mRNA | AY195839.1 | - | 3 - 4 | 342 | 79 | |
BC104567.1 | - | 3 - 4 | 399 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |