Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014864.1 | NP_001014864.1 | 1 - 2 | 121 | 58 | |
GenBank mRNA | BC102546.1 | - | 1 - 2 | 55 | 58 | |
BT020859.1 | - | 1 - 2 | 121 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |