Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014875.1 | NP_001014875.1 | 7 - 8 | 688 | 129 | |
GenBank mRNA | BC105553.1 | - | 7 - 8 | 725 | 129 | |
BT021021.1 | - | 7 - 8 | 688 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |