Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015518.1 | NP_001015518.1 | 1 - 2 | 135 | 75 | |
GenBank mRNA | BT020819.1 | - | 1 - 2 | 135 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |