Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015582.1 | NP_001015582.1 | 1 - 2 | 211 | 67 | |
GenBank mRNA | BC113270.1 | - | 1 - 2 | 190 | 67 | |
BT020651.1 | - | 1 - 2 | 203 | 67 | ||
BT020781.1 | - | 1 - 2 | 194 | 67 | ||
BT020876.1 | - | 1 - 2 | 211 | 67 | ||
BT020884.1 | - | 1 - 2 | 194 | 67 | ||
BT020898.1 | - | 1 - 2 | 195 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |