Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015626.1 | NP_001015626.1 | 2 - 3 | 255 | 94 | |
GenBank mRNA | BC103161.1 | - | 2 - 3 | 271 | 94 | |
BT021070.1 | - | 2 - 3 | 255 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |