Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT021825.1 | - | 1 - 2 | 1399 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |