Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024544.1 | NP_001019715.1 | 4 - 5 | 740 | 98 | |
GenBank mRNA | BC126704.1 | - | 5 - 6 | 835 | 98 | |
BT021834.1 | - | 4 - 5 | 740 | 98 | ||
BT026326.1 | - | 3 - 4 | 779 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |