Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024558.1 | NP_001019729.1 | 6 - 7 | 985 | 127 | |
GenBank mRNA | BC150021.1 | - | 6 - 7 | 1003 | 127 | |
BT021145.1 | - | 6 - 7 | 985 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |