Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025346.1 | NP_001020517.1 | 2 - 3 | 193 | 87 | |
GenBank mRNA | BC133652.1 | - | 2 - 3 | 205 | 87 | |
DQ062556.1 | - | 2 - 3 | 193 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |