Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025348.1 | NP_001020519.1 | 3 - 4 | 252 | 134 | |
GenBank mRNA | AY911325.1 | - | 3 - 4 | 252 | 134 | |
BC102485.1 | - | 3 - 4 | 252 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |