Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033120.1 | NP_001028292.1 | 4 - 5 | 397 | 110 | |
GenBank mRNA | AB205159.1 | - | 4 - 5 | 397 | 110 | |
BC149398.1 | - | 4 - 5 | 600 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |