Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033619.2 | NP_001028791.1 | 4 - 5 | 475 | 124 | |
GenBank mRNA | AB098916.1 | - | 4 - 5 | 450 | 123 | |
BC102624.1 | - | 4 - 5 | 476 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |