Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034327.1 | NP_001029499.1 | 1 - 2 | 403 | 69 | |
GenBank mRNA | BC120377.1 | - | 2 - 3 | 283 | 69 | |
BT021499.1 | - | 1 - 2 | 403 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |