Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034543.1 | NP_001029715.1 | 5 - 6 | 748 | 150 | |
GenBank mRNA | BC102241.1 | - | 5 - 6 | 748 | 150 | |
BT020972.1 | - | 5 - 6 | 709 | 150 | ||
BT029906.1 | - | 5 - 6 | 686 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |