Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034585.2 | NP_001029757.1 | 1 - 2 | 46 | 59 | |
GenBank mRNA | BC102992.1 | - | 1 - 2 | 47 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |