Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034643.2 | NP_001029815.1 | 5 - 6 | 706 | 73 | |
GenBank mRNA | BC102576.1 | - | 5 - 6 | 707 | 73 | |
BT029891.1 | - | 4 - 5 | 731 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |