Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035296.2 | NP_001030373.1 | 9 - 10 | 971 | 114 | |
GenBank mRNA | BC102112.1 | - | 9 - 10 | 972 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |