Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035352.1 | NP_001030429.1 | 2 - 3 | 137 | 88 | |
GenBank mRNA | BC142289.1 | - | 2 - 3 | 152 | 88 | |
BT020783.1 | - | 2 - 3 | 137 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |