Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035443.1 | NP_001030520.1 | 2 - 3 | 197 | 74 | |
GenBank mRNA | BC112734.1 | - | 2 - 3 | 254 | 74 | |
BT021492.1 | - | 2 - 3 | 197 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |