Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040543.3 | NP_001035633.1 | 3 - 4 | 267 | 80 | |
GenBank mRNA | BC103236.1 | - | 1 | 254 | 80 | |
CB426020.1 | - | 3 - 4 | 235 | 80 | ||
EV659942.1 | - | 3 - 4 | 271 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |