Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040597.1 | NP_001035687.1 | 339 | 76 | ||
GenBank mRNA | BC122799.1 | - | 5 - 6 | 359 | 76 | |
BC149205.1 | - | 1 - 2 | 1641 | 76 | ||
BT020731.1 | - | 5 - 6 | 324 | 76 | ||
BT020811.1 | - | 5 - 6 | 344 | 76 | ||
BT020814.1 | - | 5 - 6 | 330 | 76 | ||
BT020820.1 | - | 5 - 6 | 336 | 76 | ||
BT020878.1 | - | 5 - 6 | 342 | 76 | ||
BT020882.1 | - | 5 - 6 | 328 | 76 | ||
BT020897.1 | - | 5 - 6 | 331 | 76 | ||
BT020914.1 | - | 339 | 76 | |||
BT020930.1 | - | 5 - 6 | 331 | 76 | ||
BT020940.1 | - | 5 - 6 | 337 | 76 | ||
BT021643.1 | - | 5 - 6 | 333 | 76 | ||
BT021894.1 | - | 5 - 6 | 337 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |