Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040598.1 | NP_001035688.1 | 4 - 5 | 478 | 64 | |
GenBank mRNA | BC114900.1 | - | 4 - 5 | 397 | 64 | |
BT021811.1 | - | 4 - 5 | 478 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |