Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046591.2 | NP_001040056.2 | 5 - 6 | 887 | 95 | |
GenBank mRNA | BT020834.1 | - | 5 - 6 | 887 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |