Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076973.1 | NP_001070441.1 | 2 - 3 | 437 | 93 | |
GenBank mRNA | BC149326.1 | - | 2 - 3 | 433 | 93 | |
BT021776.1 | - | 2 - 3 | 437 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |