Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173926.2 | NP_776351.2 | 1 - 2 | 48 | 135 | |
GenBank mRNA | BC142034.1 | - | 1 - 2 | 41 | 135 | |
EV652245.1 | - | 1 - 2 | 94 | 135 | ||
M54979.2 | - | 1 - 2 | 25 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |