Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174116.1 | NP_776541.1 | 2 - 3 | 739 | 98 | |
GenBank mRNA | AB257294.1 | - | 2 - 3 | 684 | 98 | |
BC146221.1 | - | 2 - 3 | 726 | 98 | ||
X52526.1 | - | 649 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |