Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174164.1 | NP_776589.1 | 2 - 3 | 3244 | 76 | |
GenBank mRNA | AH001140.2 | - | 479 | 76 | ||
M27870.1 | - | 2 - 3 | 1040 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |