Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174514.2 | NP_776939.1 | 5 - 6 | 1435 | 79 | |
GenBank mRNA | AF487464.1 | - | 5 - 6 | 1435 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |