Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174686.2 | NP_777111.1 | 4 - 5 | 782 | 140 | |
GenBank mRNA | AY148474.1 | - | 4 - 5 | 782 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |