Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181016.3 | NP_851359.2 | 2 - 3 | 121 | 80 | |
GenBank mRNA | AF160867.2 | - | 2 - 3 | 141 | 80 | |
AF335552.1 | - | 1 - 2 | 86 | 80 | ||
AF540564.1 | - | 2 - 3 | 121 | 80 | ||
BC108181.1 | - | 2 - 3 | 145 | 80 | ||
KJ463389.1 | - | 1 - 2 | 86 | 80 |
Gene Symbol: M-SAA3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242573.1 | NP_001229502.1 | 2 - 3 | 145 | 80 | |
GenBank mRNA | AY943216.1 | - | 2 - 3 | 145 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |