Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033624.2 | NP_001028796.1 | 1 - 2 | 106 | 71 | |
GenBank mRNA | BC102215.1 | - | 1 - 2 | 107 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |