Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034273.2 | NP_001029445.1 | 1 - 2 | 46 | 99 | |
GenBank mRNA | BC102713.1 | - | 1 - 2 | 47 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |