Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034339.1 | NP_001029511.1 | 7 - 8 | 815 | 75 | |
GenBank mRNA | BC103219.1 | - | 7 - 8 | 815 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |