Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034491.2 | NP_001029663.1 | 6 - 7 | 845 | 65 | |
GenBank mRNA | BC102621.1 | - | 6 - 7 | 847 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |