Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034652.2 | NP_001029824.1 | 6 - 7 | 500 | 87 | |
GenBank mRNA | BC102395.1 | - | 6 - 7 | 502 | 87 | |
BT025465.1 | - | 5 - 6 | 395 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |