Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034686.2 | NP_001029858.1 | 2 - 3 | 507 | 112 | |
GenBank mRNA | BC102273.1 | - | 2 - 3 | 508 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |