Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035036.2 | NP_001030208.1 | 3 - 4 | 668 | 125 | |
GenBank mRNA | BC102165.1 | - | 3 - 4 | 669 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |