Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035293.1 | NP_001030370.1 | 6 - 7 | 1815 | 147 | |
GenBank mRNA | AY267258.1 | - | 1673 | 147 | ||
BC103349.1 | - | 6 - 7 | 1815 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |