Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035500.2 | NP_001030577.1 | 1 - 2 | 350 | 109 | |
GenBank mRNA | BC102289.1 | - | 1 - 2 | 351 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |