Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040487.2 | NP_001035577.1 | 1 - 2 | 33 | 83 | |
GenBank mRNA | BC102691.1 | - | 1 - 2 | 34 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |