Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040487.2 | NP_001035577.1 | 8 - 9 | 1059 | 69 | |
GenBank mRNA | AB098747.1 | - | 2 - 3 | 361 | 69 | |
AB098752.1 | - | 2 - 3 | 70 | |||
BC102691.1 | - | 8 - 9 | 1060 | 69 | ||
BT025451.1 | - | 7 - 8 | 1036 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |