Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076427.1 | NP_001069895.1 | 2 - 3 | 352 | 69 | |
GenBank mRNA | BC102811.1 | - | 2 - 3 | 352 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |