Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22H3orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076497.2 | NP_001069965.1 | 2 - 3 | 219 | 141 | |
GenBank mRNA | BC108183.1 | - | 2 - 3 | 220 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |