Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076998.2 | NP_001070466.1 | 3 - 4 | 178 | 136 | |
GenBank mRNA | BC103039.1 | - | 3 - 4 | 179 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |